missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Tris Base, ULTROL™ Grade, Calbiochem™,
Useful in the pH range of 10.2 to 10.6
Supplier: MilliporeSigma™ 6483115KG
Specifications
Tris Base | |
77-86-1 | |
1.353g/cm3 | |
by titration (dry basis) | |
C4H11NO3 | |
Soluble in water (800g/L at 20°C) | |
C(C(CO)(CO)N)O | |
121.136 | |
CHEBI:9754 |
168°C to 172°C | |
219°C to 220°C | |
5 kg | |
≥99.0% | |
tris(Hydroxymethyl)aminomethane | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol | |
6503 | |
121.1 |
Chemical Identifiers
77-86-1 | |
121.136 | |
tris(Hydroxymethyl)aminomethane | |
CHEBI:9754 | |
C(C(CO)(CO)N)O |
C4H11NO3 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
6503 | |
2-amino-2-(hydroxymethyl)propane-1,3-diol |
Safety and Handling
Recommended Storage : 15°C to 30°C (Do not freeze).