missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris Base (White Crystals or Crystalline Powder/Molecular Biology), Fisher BioReagents™

DNase-, RNase- and Protease-Free.

$147.00 - $4990.48

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym Trimethylol Aminomethane, Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP152-500 Edge
SDS View Documents Promotion Details
Fisher BioReagents
BP152500
500 g Poly Bottle
Each for $147.00
Case of 6 Each for $882.00
Edge
 
BP152-1
SDS View Documents Promotion Details
Fisher BioReagents
BP1521
1 kg Poly Bottle
Each for $197.25
Case of 6 Each for $1,183.50
 
BP152-5 Edge
SDS View Documents Promotion Details
Fisher BioReagents
BP1525
5 kg Poly Pail
Each for $819.00
Edge
 
BP152-10
SDS View Documents Promotion Details
Fisher BioReagents
FLBP15210
10 kg Poly Pail
Each for $1,993.48
 
BP152-25
SDS View Documents Promotion Details
Fisher BioReagents
BP15225
25 kg Fiber Drum
Each for $4,990.48
 
Description

Description

DNase-, RNase- and Protease-Free.

Tris Base is a buffer component in molecular biology, tissue culture, and electrophoresis procedures.
Specifications

Chemical Identifiers

77-86-1
121.136
Trimethylol Aminomethane, Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol
CHEBI:9754
C(C(CO)(CO)N)O
C4H11NO3
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol

Specifications

0.05 AU max. at 280nm
77-86-1
White
Crystalline Powder or Crystals
DNase free
Pass Test
C4H11NO3
Poly Bottle
15, 9951
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
121.14
DNase-, RNase- and Protease-Free
168.5°C
219°C
10.4
500 g
2-Amino-2-(Hydroxymethyl)-1,3-Propanediol, Tris (hydroxymethyl) Aminomethane, Tris Base, THAM, Trimethylol Aminomethane, Trometamol
≥99.8 %
H2NC(CH2OH)3
Protease free
Trimethylol Aminomethane, Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol
C(C(CO)(CO)N)O
121.136
CHEBI:9754
≥99.8%
Tris Base
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'