missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris(ethylenediamine)cobalt(III) chloride trihydrate, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1721109

 View more versions of this product

Catalog No. AAA1721109


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tris(ethylenediamine)cobalt(III) chloride trihydrate
C6H30Cl3CoN6O3
10 g
WBMVTRUQSMZIKW-UHFFFAOYSA-K
cobalt(3+);ethane-1,2-diamine;trichloride;trihydrate
73995044
14883-80-8
MFCD00211318
tris ethylenediamine cobalt iii chloride trihydrate, cobalt 3+ tris ethylenediamine trihydrate trichloride
C(CN)N.C(CN)N.C(CN)N.O.O.O.[Cl-].[Cl-].[Cl-].[Co+3]
399.628
399.64 (345.59 Anhydrous)

Chemical Identifiers

14883-80-8
399.628
WBMVTRUQSMZIKW-UHFFFAOYSA-K
73995044
C(CN)N.C(CN)N.C(CN)N.O.O.O.[Cl-].[Cl-].[Cl-].[Co+3]
C6H30Cl3CoN6O3
MFCD00211318
tris ethylenediamine cobalt iii chloride trihydrate, cobalt 3+ tris ethylenediamine trihydrate trichloride
cobalt(3+);ethane-1,2-diamine;trichloride;trihydrate
Safety and Handling

Safety and Handling

GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only