Learn More
Tris(ethylenediamine)cobalt(III) chloride trihydrate
CAS: 14883-80-8 | C6H30Cl3CoN6O3 | 399.628 g/mol
$106.63 - $414.85
Chemical Identifiers
| CAS | 14883-80-8 |
|---|---|
| Molecular Formula | C6H30Cl3CoN6O3 |
| Molecular Weight (g/mol) | 399.628 |
| MDL Number | MFCD00211318 |
| InChI Key | WBMVTRUQSMZIKW-UHFFFAOYSA-K |
| Synonym | tris ethylenediamine cobalt iii chloride trihydrate, cobalt 3+ tris ethylenediamine trihydrate trichloride |
| PubChem CID | 73995044 |
| IUPAC Name | cobalt(3+);ethane-1,2-diamine;trichloride;trihydrate |
| SMILES | C(CN)N.C(CN)N.C(CN)N.O.O.O.[Cl-].[Cl-].[Cl-].[Co+3] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1721109
|
Thermo Scientific Chemicals
A1721109 |
10 g |
Each for $106.63
|
|
|||||
|
AAA1721118
|
Thermo Scientific Chemicals
A1721118 |
50 g |
Each for $414.85
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 14883-80-8 | |
| 399.628 | |
| WBMVTRUQSMZIKW-UHFFFAOYSA-K | |
| 73995044 | |
| C(CN)N.C(CN)N.C(CN)N.O.O.O.[Cl-].[Cl-].[Cl-].[Co+3] |
| C6H30Cl3CoN6O3 | |
| MFCD00211318 | |
| tris ethylenediamine cobalt iii chloride trihydrate, cobalt 3+ tris ethylenediamine trihydrate trichloride | |
| cobalt(3+);ethane-1,2-diamine;trichloride;trihydrate |
Specifications
| 14883-80-8 | |
| MFCD00211318 | |
| tris ethylenediamine cobalt iii chloride trihydrate, cobalt 3+ tris ethylenediamine trihydrate trichloride | |
| C(CN)N.C(CN)N.C(CN)N.O.O.O.[Cl-].[Cl-].[Cl-].[Co+3] | |
| 399.628 | |
| 399.64 (345.59 Anhydrous) |
| C6H30Cl3CoN6O3 | |
| 10 g | |
| WBMVTRUQSMZIKW-UHFFFAOYSA-K | |
| cobalt(3+);ethane-1,2-diamine;trichloride;trihydrate | |
| 73995044 | |
| Tris(ethylenediamine)cobalt(III) chloride trihydrate |
Safety and Handling
GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only