missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
TRIS hydrochloride, >99%, Cell culture reagent grade, MP Biomedicals™
Excellent biochemical and biological buffer for all applications where ultra high purity is required. The pH values of all buffers are temperature and concentration dependent
Supplier: MP Biomedicals Inc 0219455880
Specifications
TRIS hydrochloride | |
Crystalline Powder | |
C4H12ClNO3 | |
TRIS HCl, Tris(hydroxymethyl)aminomethane hydrochloride | |
C(C(CO)(CO)N)O.Cl | |
157.594 | |
157.6 |
White | |
100 g | |
MFCD00012590 | |
QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride | |
93573 | |
Cell Culture |
Chemical Identifiers
C4H12ClNO3 | |
MFCD00012590 | |
TRIS HCl, Tris(hydroxymethyl)aminomethane hydrochloride | |
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
157.594 | |
QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
93573 | |
C(C(CO)(CO)N)O.Cl |
Safety and Handling
EINECSNumber : 214-684-5