missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TRIS hydrochloride, >99%, Cell culture reagent grade, MP Biomedicals™

Excellent biochemical and biological buffer for all applications where ultra high purity is required. The pH values of all buffers are temperature and concentration dependent

$106.21 - $247.21

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
MDL Number MFCD00012590
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym TRIS HCl, Tris(hydroxymethyl)aminomethane hydrochloride
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19455880
View Documents
MP Biomedicals Inc
0219455880
100 g
Each for $106.21
 
ICN19455890
View Documents
MP Biomedicals Inc
0219455890
500 g
Each for $247.21
 
Description

Description

Specifications

Chemical Identifiers

1185-53-1
157.594
QKNYBSVHEMOAJP-UHFFFAOYSA-N
93573
C(C(CO)(CO)N)O.Cl
C4H12ClNO3
MFCD00012590
TRIS HCl, Tris(hydroxymethyl)aminomethane hydrochloride
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride

Specifications

White
100 g
MFCD00012590
QKNYBSVHEMOAJP-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
93573
Cell Culture
Crystalline Powder
C4H12ClNO3
TRIS HCl, Tris(hydroxymethyl)aminomethane hydrochloride
C(C(CO)(CO)N)O.Cl
157.594
157.6
TRIS hydrochloride
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-684-5

SDS
missing translation for 'documents'

missing translation for 'documents'