missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris(pentafluorophenyl)borane, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 353130010

 View more versions of this product

Catalog No. AC353130010

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tris(pentafluorophenyl)borane
1109-15-5
94% min.
94% min
C18BF15
MFCD00269813
15, 9931
OBAJXDYVZBHCGT-UHFFFAOYSA-N
tris(2,3,4,5,6-pentafluorophenyl)borane
582056
95%
95%
126°C to 131°C
Authentic
Glass bottle
(C6F5)3B
1 g
tris pentafluorophenyl borane, tris perfluorophenyl borane, tris pentafluorophenyl boron, tris 2,3,4,5,6-pentafluorophenyl borane, unii-i3wu5e2578, tris pentafluorophenyl-borane, borane, tris pentafluorophenyl, borane, tris 2,3,4,5,6-pentafluorophenyl, perfluorotriphenylboron, zlchem 1272
FC1=C(F)C(F)=C(B(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
511.98
511.99
Powder

Chemical Identifiers

1109-15-5
511.98
OBAJXDYVZBHCGT-UHFFFAOYSA-N
582056
FC1=C(F)C(F)=C(B(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
C18BF15
MFCD00269813
tris pentafluorophenyl borane, tris perfluorophenyl borane, tris pentafluorophenyl boron, tris 2,3,4,5,6-pentafluorophenyl borane, unii-i3wu5e2578, tris pentafluorophenyl-borane, borane, tris pentafluorophenyl, borane, tris 2,3,4,5,6-pentafluorophenyl, perfluorotriphenylboron, zlchem 1272
tris(2,3,4,5,6-pentafluorophenyl)borane
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap a

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only