Troemner™ Analytical Weight Cases
Polycarbonate cases securely hold and protect ASTM weights of various sizes
Description
- Designed to hold and protect weights sized 2kg through 10g
- Lid is rotated open, color-coded insert and weight move with lid
- Prevents urethane bumper from rubbing on weight, further protecting weight's calibration
Specifications
Individual Analytical Weight Case | |
3.5 in. | |
500g weight |
Polycarbonate | |
4.5 in. | |
Each |