Learn More
Troemner™ Analytical Weight Cases
Polycarbonate cases securely hold and protect ASTM weights of various sizes
$67.67 - $181.03
Specifications
| Product Type | Individual Analytical Weight Case |
|---|---|
| Material | Polycarbonate |
| Quantity | Each |
| Catalog Number | Mfr. No. | Diameter (English) Outside | For Use With (Equipment) | Height (English) Case | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (English) Outside | For Use With (Equipment) | Height (English) Case | Price | Quantity | |||
|
02-215-546
|
Troemner™
80864210 |
2 in. | 7mg weight | 2.5 in. |
Each for $70.74
|
|
|||
|
02-215-547
|
Troemner™
80864211 |
2 in. | 1g weight | 2.5 in. |
Each for $67.67
|
|
|||
|
02-215-548
|
Troemner™
80864212 |
2 in. | 2g weight | 2.5 in. |
Each for $92.52
|
|
|||
|
02-215-550
|
Troemner™
80864214 |
2 in. | 5g weight | 2.5 in. |
Each for $67.67
|
|
|||
|
02-215-551
|
Troemner™
80864215 |
2 in. | 10g weight | 2.5 in. |
Each for $70.74
|
|
|||
|
02-215-552
|
Troemner™
80864216 |
2 in. | 20g weight | 2.5 in. |
Each for $67.67
|
|
|||
|
02-215-554
|
Troemner™
80864218 |
2 in. | 50g weight | 2.5 in. |
Each for $76.09
|
|
|||
|
02-215-555
|
Troemner™
80864219 |
2 in. | 100g weight | 2.5 in. |
Each for $71.96
|
|
|||
|
02-215-556
|
Troemner™
80864220 |
2 in. | 200g weight | 2.5 in. |
Each for $79.52
|
|
|||
|
02-215-553
|
Troemner™
80864217 |
2 in. | 30g weight | 2.5 in. |
Each for $70.74
|
|
|||
|
02-215-557
|
Troemner™
80864221 |
2 in. | 300g weight | 2.5 in. |
Each for $104.09
|
|
|||
|
02-215-558
|
N/A | 3.5 in. | 500g weight | 4.5 in. |
Each for $172.77
|
|
|||
|
02-215-559
|
N/A | 3.5 in. | 1kg weight | 4.5 in. |
Each for $181.03
|
|
|||
|
02-215-560
|
Troemner™
80864225 |
3.5 in. | 2kg weight | 4.5 in. |
Each for $110.63
|
|
|||
Description
- Designed to hold and protect weights sized 2kg through 10g
- Lid is rotated open, color-coded insert and weight move with lid
- Prevents urethane bumper from rubbing on weight, further protecting weight's calibration
Specifications
| Individual Analytical Weight Case | |
| Polycarbonate | |
| Each |