Troemner™ Cast Iron Support Stand Bases
Cast iron support stand base
Supplier:  Troemner™ 30392363
 View more versions of this product
Catalog No. 02-300-242
Explore available promotions
Description
Description
  • Constructed with a black enamel finish for chemical resistance and durability
  • Base features a built-in support rod holder with locking knob that accepts a 0.51 in. (13mm) dia. support rod
  • Support rods are not included
Specifications
Specifications
Cast Iron
0.51 in.
Support stand base only
Black Enamel
Each
14.75 in.
15.2 cm
Tripod
Standard and heavy duty support stand
Videos
SDS
Documents
Documents
Accessories