Troemner™ Metric Economical Stainless Steel Calibration Weight Sets, Class 7
Economical, quality weights
 View more versions of this product
Catalog No. 02301103
Explore available promotions
Description
Description
  • Sets designed to achieve any total mass by combining smaller masses
  • Weights 1g to 100g are of one-piece construction and are adjusted during manufacture by removing material from the bottom of the weight
  • Weights 200g through 2kg have an adjusting cavity on the top that is sealed with a plug
Compliance
Adjusted to tolerances specified by ANSI/ASTM E617 Class 7
Specifications
Specifications
Metric Economical Weight Set
Stainless Steel
7.85g/cm3 at 20°C
Cylindrical with Handling Knob
12/Set
ASTM Class 7
1 to 500 g
1 g to 500 g
Cylindrical weight: 500g, 2 x 200g, 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Plastic case
Multiple