Troemner™ Metric Economical Stainless Steel Calibration Weight Sets, Class 7
Economical, quality weights
 View more versions of this product
Catalog No. 2301105
Explore available promotions
Description
Description
  • Sets designed to achieve any total mass by combining smaller masses
  • Weights 1g to 100g are of one-piece construction and are adjusted during manufacture by removing material from the bottom of the weight
  • Weights 200g through 2kg have an adjusting cavity on the top that is sealed with a plug
Compliance
Adjusted to tolerances specified by ANSI/ASTM E617 Class 7
Specifications
Specifications
Metric Economical Weight Set
Stainless Steel
7.85g/cm3 at 20°C
Sheet
12/Set
ASTM Class 7
1 mg to 500 g
1 mg to 500 mg
Sheet weight: 500mg, 2 x 200mg, Aluminum sheet weight: 100mg, 50mg, 2 x 20mg, 10mg, 5mg, 2 x 2mg, 1mg, Plastic case
Multiple