Troemner™ Stainless Steel Electronic Balance Calibration Weights, Class 4
Precision-crafted of stainless steel, with a highly polished finish
 View more versions of this product
Catalog No. 02-215-65K
Explore available promotions
Description
Description
All electronic balance weights are manufactured with adjusting cavities
  • Weights 5kg to 30kg are flat on top with a recessed grip handle for easy handling and stacking
  • Weights 1kg to 5kg are cylindrical in shape with a groove near the top to allow for easy handling and gripping
  • Weights below 1kg are straight cylinders
  • Fractional weights 500 mg to 5 mg are made of stainless steel (30 mg weights are aluminum); fractional weights 3 mg and below are made of aluminum
  • Weights 5kg and below supplied in a rugged, precision-fit, high quality polycarbonate case with handling glove
Specifications
Specifications
Two-Piece Electronic Balance Calibration Weight
Stainless Steel
7.95g/cm3 at 20°C
Straight Cylinder
Each
ASTM Class 4
500 g
500 g
Polypropylene case
1