Fisherbrand™ TRUFLOW™ Tissue and Biopsy Cassettes
Just bend, snap apart, and conveniently lock
 View more versions of this product
Catalog No. 15-200-403E
Explore available promotions
Description
Description
  • Hold tissue and biopsy specimens during the embedding process as well as in a storage file
  • Molded from a special high-density polymer that is totally resistant to histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • One-piece integral lid eliminates the need for separate steel lids
  • Can be opened or closed as often as necessary and relocked securely without danger of specimen loss
  • Large labeling areas are located on three sides of the cassette
  • Anterior writing area at a 30° angle
Order Info
Packaged in three dispenser boxes of 500
Specifications
Specifications
Slots
Integral Lid
High Density Polymer
30°
White
Tissue Cassette
Product Suggestions
Product Suggestions