missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tryptamine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 186120100

 View more versions of this product

Catalog No. AC186120100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tryptamine
113.0°C to 117.0°C
137.0°C (0.1 mmHg)
Authentic
Glass bottle
MFCD00005661
22,I,636
tryptamine, 2-1h-indol-3-yl ethanamine, 3-2-aminoethyl indole, 1h-indole-3-ethanamine, 2-3-indolyl ethylamine, indol-3-ethylamine, indole, 3-2-aminoethyl, 2-indol-3-yl ethylamine, 2-1h-indol-3-yl ethan-1-amine, tryptamin
APJYDQYYACXCRM-UHFFFAOYSA-N
2-(1H-indol-3-yl)ethanamine
1150
160.22
Crystalline Powder or Flakes
61-54-1
White
185°C
97.5% min. (GC)
C10H12N2
10 g
15,9976
Solubility in water: negligible. Other solubilities: soluble in ethanol and acetone, practically insoluble in ether, benzene and, chloroform
NCCC1=CNC2=CC=CC=C12
160.22
CHEBI:16765
98%

Chemical Identifiers

61-54-1
160.22
APJYDQYYACXCRM-UHFFFAOYSA-N
1150
2-(1H-indol-3-yl)ethanamine
C10H12N2
MFCD00005661
tryptamine, 2-1h-indol-3-yl ethanamine, 3-2-aminoethyl indole, 1h-indole-3-ethanamine, 2-3-indolyl ethylamine, indol-3-ethylamine, indole, 3-2-aminoethyl, 2-indol-3-yl ethylamine, 2-1h-indol-3-yl ethan-1-amine, tryptamin
CHEBI:16765
NCCC1=CNC2=CC=CC=C12
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-510-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only