missing translation for 'onlineSavingsMsg'
Learn More

Tryptamine, 98%

CAS: 61-54-1 | C10H12N2 | 160.22 g/mol

$82.86 - $266.12

Chemical Identifiers

CAS 61-54-1
Molecular Formula C10H12N2
Molecular Weight (g/mol) 160.22
MDL Number MFCD00005661
InChI Key APJYDQYYACXCRM-UHFFFAOYSA-N
Synonym tryptamine, 2-1h-indol-3-yl ethanamine, 3-2-aminoethyl indole, 1h-indole-3-ethanamine, 2-3-indolyl ethylamine, indol-3-ethylamine, indole, 3-2-aminoethyl, 2-indol-3-yl ethylamine, 2-1h-indol-3-yl ethan-1-amine, tryptamin
PubChem CID 1150
ChEBI CHEBI:16765
IUPAC Name 2-(1H-indol-3-yl)ethanamine
SMILES NCCC1=CNC2=CC=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC186120100
SDS View Documents
Thermo Scientific Chemicals
186120100
10 g Glass bottle
Each for $82.86
Only null left
 
AC186120500
SDS View Documents
Thermo Scientific Chemicals
186120500
50 g Glass bottle
Each for $266.12
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

61-54-1
160.22
APJYDQYYACXCRM-UHFFFAOYSA-N
1150
2-(1H-indol-3-yl)ethanamine
C10H12N2
MFCD00005661
tryptamine, 2-1h-indol-3-yl ethanamine, 3-2-aminoethyl indole, 1h-indole-3-ethanamine, 2-3-indolyl ethylamine, indol-3-ethylamine, indole, 3-2-aminoethyl, 2-indol-3-yl ethylamine, 2-1h-indol-3-yl ethan-1-amine, tryptamin
CHEBI:16765
NCCC1=CNC2=CC=CC=C12

Specifications

61-54-1
White
185°C
97.5% min. (GC)
C10H12N2
10 g
15,9976
Solubility in water: negligible. Other solubilities: soluble in ethanol and acetone, practically insoluble in ether, benzene and, chloroform
NCCC1=CNC2=CC=CC=C12
160.22
CHEBI:16765
98%
Tryptamine
113.0°C to 117.0°C
137.0°C (0.1 mmHg)
Authentic
Glass bottle
MFCD00005661
22,I,636
tryptamine, 2-1h-indol-3-yl ethanamine, 3-2-aminoethyl indole, 1h-indole-3-ethanamine, 2-3-indolyl ethylamine, indol-3-ethylamine, indole, 3-2-aminoethyl, 2-indol-3-yl ethylamine, 2-1h-indol-3-yl ethan-1-amine, tryptamin
APJYDQYYACXCRM-UHFFFAOYSA-N
2-(1H-indol-3-yl)ethanamine
1150
160.22
Crystalline Powder or Flakes
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-510-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only