missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten(VI) chloride, 99.9+%, (trace metal basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 221430100

 View more versions of this product

Catalog No. AC221430100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
Specifications

Specifications

347°C
275°C
Blue-Gray to Gray
10 g
Cl6W
MFCD00011463
tungsten vi chloride, tungsten hexachloride, wolfram hexachloride, wcl6, tungsten chloride wcl6 , oc-6-11, tungsten chloride, oc-6-11, tungsten chloride 8ci, tungsten 6+ hexachloride, wolframhexachlorid, hexachloridotungsten
KPGXUAIFQMJJFB-UHFFFAOYSA-H
hexachlorotungsten
83301
396.57
Trace Metal Basis
Tungsten(VI) chloride
13283-01-7
Powder
99.9+%
WCl6
04,569; 05,742; 06,658; 14,346
Solubility in water: decomposes. Other solubilities: soluble in cs2,chloroform,methyl alcohol,ether,,acetone,pyridine,glycerol,ligroin,ethanol,,carbon tetrachloride,phosphorus oxytrichloride,,benzene
Cl[W](Cl)(Cl)(Cl)(Cl)Cl
396.57
CHEBI:37771
≥99.9%
Glass bottle

Chemical Identifiers

13283-01-7
396.57
KPGXUAIFQMJJFB-UHFFFAOYSA-H
83301
hexachlorotungsten
Cl6W
MFCD00011463
tungsten vi chloride, tungsten hexachloride, wolfram hexachloride, wcl6, tungsten chloride wcl6 , oc-6-11, tungsten chloride, oc-6-11, tungsten chloride 8ci, tungsten 6+ hexachloride, wolframhexachlorid, hexachloridotungsten
CHEBI:37771
Cl[W](Cl)(Cl)(Cl)(Cl)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 236-293-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only