Learn More
Tungsten(VI) chloride, 99.9+%, (trace metal basis), Thermo Scientific Chemicals
$105.30 - $496.11
Chemical Identifiers
CAS | 13283-01-7 |
---|---|
Molecular Formula | Cl6W |
Molecular Weight (g/mol) | 396.57 |
MDL Number | MFCD00011463 |
InChI Key | KPGXUAIFQMJJFB-UHFFFAOYSA-H |
Synonym | tungsten vi chloride, tungsten hexachloride, wolfram hexachloride, wcl6, tungsten chloride wcl6 , oc-6-11, tungsten chloride, oc-6-11, tungsten chloride 8ci, tungsten 6+ hexachloride, wolframhexachlorid, hexachloridotungsten |
PubChem CID | 83301 |
ChEBI | CHEBI:37771 |
IUPAC Name | hexachlorotungsten |
SMILES | Cl[W](Cl)(Cl)(Cl)(Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC221430100
|
Thermo Scientific Chemicals
221430100 |
10 g | Glass bottle |
Each for $105.30
|
|
|||||
AC221431000
|
Thermo Scientific Chemicals
221431000 |
100 g | Glass bottle |
Each for $496.11
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisChemical Identifiers
13283-01-7 | |
396.57 | |
KPGXUAIFQMJJFB-UHFFFAOYSA-H | |
83301 | |
hexachlorotungsten |
Cl6W | |
MFCD00011463 | |
tungsten vi chloride, tungsten hexachloride, wolfram hexachloride, wcl6, tungsten chloride wcl6 , oc-6-11, tungsten chloride, oc-6-11, tungsten chloride 8ci, tungsten 6+ hexachloride, wolframhexachlorid, hexachloridotungsten | |
CHEBI:37771 | |
Cl[W](Cl)(Cl)(Cl)(Cl)Cl |
Specifications
347°C | |
13283-01-7 | |
Powder | |
99.9+% | |
WCl6 | |
04,569; 05,742; 06,658; 14,346 | |
Solubility in water: decomposes. Other solubilities: soluble in cs2,chloroform,methyl alcohol,ether,,acetone,pyridine,glycerol,ligroin,ethanol,,carbon tetrachloride,phosphorus oxytrichloride,,benzene | |
Cl[W](Cl)(Cl)(Cl)(Cl)Cl | |
396.57 | |
CHEBI:37771 | |
≥99.9% | |
Glass bottle |
275°C | |
Blue-Gray to Gray | |
10 g | |
Cl6W | |
MFCD00011463 | |
tungsten vi chloride, tungsten hexachloride, wolfram hexachloride, wcl6, tungsten chloride wcl6 , oc-6-11, tungsten chloride, oc-6-11, tungsten chloride 8ci, tungsten 6+ hexachloride, wolframhexachlorid, hexachloridotungsten | |
KPGXUAIFQMJJFB-UHFFFAOYSA-H | |
hexachlorotungsten | |
83301 | |
396.57 | |
Trace Metal Basis | |
Tungsten(VI) chloride |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 236-293-9
RUO – Research Use Only