DuPont™ Tychem™ 4000 Series 122 Coveralls
Tychem™ 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges.
Supplier:  DuPont™ SL122BWH2X001200
 View more versions of this product
Catalog No. 18-999-2216
Explore available promotions
Description
Description
Tychem™ 4000 delivers effective protection against a range of chemical environments. Tychem™ 4000, utilizes Saranex™ 23-P film laminated to Tyvek™ fabric to provide a rugged and durable fabric that resists abrasion. Tychem™ 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments.
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Attached hood with elastic around face opening
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
Compliance
Meets ASTM™ D3776, ASTM D1777, ASTM D3786, ASTM D5034 standards.
Specifications
Specifications
Hooded Coveralls with Socks
Tyvek with Saranex 23-P Film Laminate
Single-use
Elastic
Attached, Elastic Face
Bound
Attached hood, elastic wrist, attached socks
30 in.
12 mil
38.25 in.
28.25 in.
6.08 to 6.33 ft.
2X-Large
White
Attached Socks
Hooded
Front Zipper
Non-sterile
44.25 to 47.75 in.
76.2 cm
Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
97.15 cm
71.75 cm
1.82 to 1.92 m