missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tychem™ 4000 Series 125 Coveralls
Useful when handling many wet and dry chemicals, and certain hazardous materials such as PCBs - See more at: http://beta.fishersci.com/shop/products//p-25013#sthash.pbR5bBoS.dpuf
Supplier: DuPont™ SL125BWHMD001200
Description
Effective protection against a range of chemicals. Excellent for waste management, PCB clean-up, hazardous response, and nuclear environments. - See more at: http://beta.fishersci.com/shop/products//p-25013#sthash.pbR5bBoS.dpuf
- Bound-Seamed models are tightly sewn with a reinforced outer binding to increase strength and particle barrier
- Taped-Seamed models are sewn and taped to provide chemical resistance against heavy liquid splashes and tough seam stress
- Laydown collar
- Front zipper closure
- Storm flap with tape closure
- Eastic wrists
- Elastic ankles
Compliance
Meets ASTM™ D3776, ASTM D1777, ASTM D3786, ASTM D5034 standards.
Specifications
Medium | |
White | |
Elastic | |
Front Zipper | |
Elastic wrist and ankle, collar; Tyvek™ protective Fabric laminated with chemical-resistant Dow Saranex™ 23-P film | |
89.53 to 98.42 cm | |
33.75 in. | |
24.25 in. | |
1.6 to 1.73 m |
Tychem | |
Elastic | |
Collared | |
Bound | |
27.5 in. | |
Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments | |
85.72 cm | |
5.3 to 5.7 ft. |