DuPont™ Tychem™ 4000 Series 125 Coveralls
Useful when handling many wet and dry chemicals, and certain hazardous materials such as PCBs - See more at: http://beta.fishersci.com/shop/products//p-25013#sthash.pbR5bBoS.dpuf
$302.25 - $407.98
Specifications
Material Tychem
Color White
Closure Type Front Zipper
View More Specs
Products 11
Catalog Number Mfr. No. Seam Type Size Price Quantity  
Catalog Number Mfr. No. Seam Type Size Price Quantity  
19-162-778
View Documents
DuPont™
SL125TWH5X000600
Taped 5X-Large N/A
19-715-335
View Documents
DuPont™
SL125TWHLG000600
Taped Large N/A
18-999-2583
View Documents
DuPont™
SL125BWH4X001200
Bound 4X-Large N/A
19-715-334
View Documents
DuPont™
SL125TWHMD000600
Taped Medium N/A
18-760D
View Documents
DuPont™
SL125BWHXL001200
Bound X-Large N/A
18-760E
View Documents
DuPont™
SL125BWH3X001200
Bound 3X-Large N/A
19-162-777
View Documents
DuPont™
SL125TWH4X000600
Taped 4X-Large N/A
19-715-336
View Documents
DuPont™
SL125TWH2X000600
Taped 2X-Large N/A
18-760C
View Documents
DuPont™
SL125BWHLG001200
Bound Large N/A
19-087309
View Documents
DuPont™
SL125BWH2X001200
Bound 2X-Large N/A
18-760B
View Documents
DuPont™
SL125BWHMD001200
Bound Medium N/A
Description
Description
Effective protection against a range of chemicals. Excellent for waste management, PCB clean-up, hazardous response, and nuclear environments. - See more at: http://beta.fishersci.com/shop/products//p-25013#sthash.pbR5bBoS.dpuf
  • Bound-Seamed models are tightly sewn with a reinforced outer binding to increase strength and particle barrier
  • Taped-Seamed models are sewn and taped to provide chemical resistance against heavy liquid splashes and tough seam stress
  • Laydown collar
  • Front zipper closure
  • Storm flap with tape closure
  • Eastic wrists
  • Elastic ankles
Compliance
Meets ASTM™ D3776, ASTM D1777, ASTM D3786, ASTM D5034 standards.
Specifications
Specifications
Tychem
White
Front Zipper
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'