DuPont™ Tychem™ 4000 Series 125 Coveralls
Useful when handling many wet and dry chemicals, and certain hazardous materials such as PCBs - See more at: http://beta.fishersci.com/shop/products//p-25013#sthash.pbR5bBoS.dpuf
Supplier:  DuPont™ SL125TWH5X000600
 View more versions of this product
Catalog No. 19-162-778
Explore available promotions
Description
Description
Effective protection against a range of chemicals. Excellent for waste management, PCB clean-up, hazardous response, and nuclear environments. - See more at: http://beta.fishersci.com/shop/products//p-25013#sthash.pbR5bBoS.dpuf
  • Bound-Seamed models are tightly sewn with a reinforced outer binding to increase strength and particle barrier
  • Taped-Seamed models are sewn and taped to provide chemical resistance against heavy liquid splashes and tough seam stress
  • Laydown collar
  • Front zipper closure
  • Storm flap with tape closure
  • Eastic wrists
  • Elastic ankles
Compliance
Meets ASTM™ D3776, ASTM D1777, ASTM D3786, ASTM D5034 standards.
Specifications
Specifications
5X-Large
White
Taped
Waste management, hazardous response and nuclear,environments
Tychem
Front Zipper
Tyvek™ protective Fabric laminated with chemical-resistant Dow Saranex™ 23-P film