missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tyramine hydrochloride, 99%, Thermo Scientific Chemicals

Tyramine hydrochloride, CAS # 60-19-5, is the hydrochloride salt of tyramine, a biogenic amine derived from the amino acid tyrosine.

Supplier:  Thermo Scientific Chemicals 422380050

 View more versions of this product

Catalog No. AC422380050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Tyramine is a naturally occurring monoamine that is derived from the decarboxylation of the amino acid tyrosine utilized in human and invertebrate nervous systems
  • Tyramine is synthesized and released from neurons and acts as a neuromodulator upon binding to tyramine receptors
  • Tyramine has been found to impact the immune system and cardiovascular system in humans

Applications

  • Tyramine may be used to form tyramide conjugate, which can be used as a signal amplification reagent in immunofluorescence assays
  • Tyramine-hyaluronic acid conjugates can be used to form biodegradable hydrogels, with utility in drug delivery and tissue engineering
  • Tyramine is useful for laboratory studies of neuronal stimulation
Specifications

Specifications

Tyramine hydrochloride
271°C to 274°C
269°C
Authentic
Glass bottle
HOC6H4CH2CH2NH2·HCl
5 g
15, 10016
(10% in water) Clear colorless to light yellow
[H+].[Cl-].NCCC1=CC=C(O)C=C1
173.64
173.65
Crystalline Powder
60-19-5
White
0.5% max. (105°C, 2 hrs)
98.5 to 101.5% (on dry substance) (Argentometry)
C8H12ClNO
MFCD00012901
13, 625
tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147
RNISDHSYKZAWOK-UHFFFAOYSA-N
hydrogen 4-(2-aminoethyl)phenol chloride
66449
99%

Chemical Identifiers

60-19-5
173.64
RNISDHSYKZAWOK-UHFFFAOYSA-N
66449
C8H12ClNO
MFCD00012901
tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147
[H+].[Cl-].NCCC1=CC=C(O)C=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 200-462-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only