missing translation for 'onlineSavingsMsg'
Learn More

Tyramine hydrochloride, 99%

Tyramine hydrochloride, CAS # 60-19-5, is the hydrochloride salt of tyramine, a biogenic amine derived from the amino acid tyrosine. | CAS: 60-19-5 | C8H12ClNO | 173.64 g/mol

$91.90 - $186.93

Chemical Identifiers

CAS 60-19-5
Molecular Formula C8H12ClNO
Molecular Weight (g/mol) 173.64
MDL Number MFCD00012901
InChI Key RNISDHSYKZAWOK-UHFFFAOYSA-N
Synonym tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147
PubChem CID 66449
SMILES [H+].[Cl-].NCCC1=CC=C(O)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC422380050
SDS View Documents
Thermo Scientific Chemicals
422380050
5 g Glass bottle
Each for $91.90
Only null left
 
AC422380250
SDS View Documents
Thermo Scientific Chemicals
422380250
25 g Glass bottle
Each for $186.93
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Tyramine is a naturally occurring monoamine that is derived from the decarboxylation of the amino acid tyrosine utilized in human and invertebrate nervous systems
  • Tyramine is synthesized and released from neurons and acts as a neuromodulator upon binding to tyramine receptors
  • Tyramine has been found to impact the immune system and cardiovascular system in humans

Applications

  • Tyramine may be used to form tyramide conjugate, which can be used as a signal amplification reagent in immunofluorescence assays
  • Tyramine-hyaluronic acid conjugates can be used to form biodegradable hydrogels, with utility in drug delivery and tissue engineering
  • Tyramine is useful for laboratory studies of neuronal stimulation
Specifications

Chemical Identifiers

60-19-5
173.64
RNISDHSYKZAWOK-UHFFFAOYSA-N
66449
C8H12ClNO
MFCD00012901
tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147
[H+].[Cl-].NCCC1=CC=C(O)C=C1

Specifications

60-19-5
White
0.5% max. (105°C, 2 hrs)
98.5 to 101.5% (on dry substance) (Argentometry)
C8H12ClNO
MFCD00012901
13, 625
tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147
RNISDHSYKZAWOK-UHFFFAOYSA-N
hydrogen 4-(2-aminoethyl)phenol chloride
66449
99%
Tyramine hydrochloride
271°C to 274°C
269°C
Authentic
Glass bottle
HOC6H4CH2CH2NH2·HCl
5 g
15, 10016
(10% in water) Clear colorless to light yellow
[H+].[Cl-].NCCC1=CC=C(O)C=C1
173.64
173.65
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 200-462-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only