Learn More
Tyramine hydrochloride, 99%, Thermo Scientific Chemicals
Tyramine hydrochloride, CAS # 60-19-5, is the hydrochloride salt of tyramine, a biogenic amine derived from the amino acid tyrosine.
Supplier: Thermo Scientific Chemicals 422380250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tyramine hydrochloride | |
271°C to 274°C | |
269°C | |
Authentic | |
Glass bottle | |
HOC6H4CH2CH2NH2·HCl | |
25 g | |
15, 10016 | |
(10% in water) Clear colorless to light yellow | |
[H+].[Cl-].NCCC1=CC=C(O)C=C1 | |
173.64 | |
173.65 | |
Crystalline Powder |
60-19-5 | |
White | |
0.5% max. (105°C, 2 hrs) | |
98.5 to 101.5% (on dry substance) (Argentometry) | |
C8H12ClNO | |
MFCD00012901 | |
13, 625 | |
tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147 | |
RNISDHSYKZAWOK-UHFFFAOYSA-N | |
4-(2-aminoethyl)phenol;hydrochloride | |
66449 | |
99% |
Chemical Identifiers
60-19-5 | |
173.64 | |
RNISDHSYKZAWOK-UHFFFAOYSA-N | |
66449 |
C8H12ClNO | |
MFCD00012901 | |
tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147 | |
[H+].[Cl-].NCCC1=CC=C(O)C=C1 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 200-462-5
RUO – Research Use Only