DuPont™ Tyvek™ 400 Frocks, Open Wrists
Tyvek frock features a body-length jacket design with front snap closures for easy donning and doffing. Available in individually packaged styles for PPE vending machines.
Supplier:  DuPont™ TY210SWH3X0030VP
 View more versions of this product
Catalog No. 19-820-538
Explore available promotions
Description
Description
Tyvek frock features a body-length jacket design with front snap closures for easy donning and doffing. Available in individually packaged styles for PPE vending machines.
  • Flash spun high density polyethylene nonwoven material
  • Provides ideal balance of protection, durability, and comfort
  • Offers an inherent barrier against particles (down to 1.0 micron in size)
  • Common applications include lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up
  • Frocks provide body-length jacket design with front snap closures for easy donning and doffing
  • Frock extends below hip
  • Laydown collar
Specifications
Specifications
General Purpose Frock
Tyvek
Limited-use
Open
Lead and asbestos abatement and remediation, general maintenance and operations, spray painting, general clean-up
Below Hip-length
ASTM D1777
44.5 in.
3X-Large
White
Snaps
Serged
Laydown
48.75 to 52.25 in.
Individually Packaged for Vending
113.03 cm