DuPont™ Tyvek™ 400 Lab Coats, USDA, FDA, OSHA
Limited-use Tyvek 400 lab coats provide protection and durability. Meets USDA, FDA, OSHA Standards.
Supplier:  DuPont™ TY212SWH4X0008G1
 View more versions of this product
Catalog No. 17-500-268
Explore available promotions
Description
Description
DuPont™ Tyvek™ 400 provides lightweight inherent barrier protection against hazardous dry particles and aerosols, and nonhazardous light liquid splash.
  • Excellent abrasion resistance due its unique, nonwoven material
  • Can be used in a variety of applications including general maintenance/operations, asbestos and lead abatement, mold remediation, and environmental cleanups
Compliance
USDA, FDA, OSHA
Specifications
Specifications
General Purpose Lab Coat
Unisex
Snaps
Non-sterile
USDA, FDA, OSHA
4X-Large
White
Open
Plain