DuPont™ Tyvek™ 400 Lab Coats, USDA, FDA, OSHA
Limited-use Tyvek 400 lab coats provide protection and durability. Meets USDA, FDA, OSHA Standards.
$61.03 - $74.01
Specifications
Product Type General Purpose Lab Coat
Gender Unisex
Color White
Closure Type Snaps
Cuff Style Open
View More Specs
Products 6
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-500-264
View Documents
DuPont™
TY212SWH2X0008G1
2X-Large
Case of 8 Each for $64.66
17-500-266
View Documents
DuPont™
TY212SWH3X0008G1
3X-Large
Case of 8 Each for $68.63
17-500-268
View Documents
DuPont™
TY212SWH4X0008G1
4X-Large
Case of 8 Each for $74.01
17-500-273
View Documents
DuPont™
TY212SWHLG0008G1
Large
Case of 8 Each for $61.03
17-500-275
View Documents
DuPont™
TY212SWHMD0008G1
Medium
Case of 8 Each for $61.03
17-500-278
View Documents
DuPont™
TY212SWHXL0008G1
X-Large
Case of 8 Each for $61.03
Description
Description
DuPont™ Tyvek™ 400 provides lightweight inherent barrier protection against hazardous dry particles and aerosols, and nonhazardous light liquid splash.
  • Excellent abrasion resistance due its unique, nonwoven material
  • Can be used in a variety of applications including general maintenance/operations, asbestos and lead abatement, mold remediation, and environmental cleanups
Compliance
USDA, FDA, OSHA
Specifications
Specifications
General Purpose Lab Coat
White
Open
Plain
Unisex
Snaps
Non-sterile
USDA, FDA, OSHA
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'