DuPont™ Tyvek™ 400 Lab Coats, USDA, FDA, OSHA
Limited-use Tyvek 400 lab coats provide protection and durability. Meets USDA, FDA, OSHA Standards.
Supplier: DuPont™ TY212SWHLG0008G1
Description
DuPont™ Tyvek™ 400 provides lightweight inherent barrier protection against hazardous dry particles and aerosols, and nonhazardous light liquid splash.
- Excellent abrasion resistance due its unique, nonwoven material
- Can be used in a variety of applications including general maintenance/operations, asbestos and lead abatement, mold remediation, and environmental cleanups
Compliance
USDA, FDA, OSHA
Specifications
General Purpose Lab Coat | |
Unisex | |
Snaps | |
Non-sterile | |
USDA, FDA, OSHA |
Large | |
White | |
Open | |
Plain |