DuPont™ Tyvek™ 400 Hooded Coveralls with Skid-Resistant Boots, Vending Packaged
Coveralls with attached skid-resistant boots include stormflaps and a respirator fit hood. Individually packaged for PPE vending machines.
Supplier:  DuPont™ TY122SWH4X0025VP
 View more versions of this product
Catalog No. 19-820-512
Explore available promotions
Description
Description
  • Comfort fit design based on extensive wearer input, individually packaged for PPE vending machine use
  • Enables a greater range of movement while stretching and bending
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Longer zipper extends to chin for complete coverage of neck area
  • Elastic opening for tighter fit at wrist
  • Attached Tyvek™ boots have special coating to provide added skid-resistance
  • 25 Coveralls/Cs.
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic
Elastic
Elastic, Respirator Fit
Serged
Particle
Lead and Asbestos Abatement, Remediation, General Maintenance and Operations, Spray Painting, General Cleaning
4X-Large
Tyvek
Limited-use
Elastic, Attached Boots
Hooded
Front Zipper
Non-sterile
TY122SWH
Individually Packaged for PPE Vending
Product Suggestions
Product Suggestions