missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tyvek™ 600 Coverall with Hood, Elastic Ankles, Thumbloops
DuPont™ Tyvek™ 600 Coverall with Hood provides an effective barrier against low concentration water-based inorganic chemicals, small sized hazardous particles down to one micron, and biological hazards.
Supplier: DuPont™ TY198TWH2X0025PI
Description
TYVEK™ 600 Coverall coveralls are ideal for occupations that require effective whole suit protection offered by a Type 4 suit because they are exposed to heavy liquid aerosols or toxic airborne solid particulates.
Enhanced design for increased flexibility and comfort:
- Improved garment fit: extra flexibility and freedom of movement
- New hood design: optimized to fit the contours of the face, ensuring fewer leakage points during head movement
- Larger zipper puller: much easier to grasp when wearing gloves.
- Certified to latest European Standards: including the Type 4 spray test.
Certifications
Chemical Protective Clothing, Cat.III,Type 5-B,6-B
Specifications
2X-Large | |
Tyvek | |
Single-use | |
Elastic | |
Taped | |
Liquid, Particle | |
Packaged Individually | |
None |
Unisex | |
White | |
Elastic | |
Hooded | |
Non-sterile | |
Chemical Protective Clothing, Cat.III,Type 5-B,6-B | |
Yes | |
25/Cs. |