DuPont™ Tyvek™ 600 Coverall with Hood, Elastic Ankles, Thumbloops
DuPont™ Tyvek™ 600 Coverall with Hood provides an effective barrier against low concentration water-based inorganic chemicals, small sized hazardous particles down to one micron, and biological hazards.
$322.36 - $492.93
Specifications
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Ankle Style Elastic
View More Specs
Products 9
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
18-888-822
View Documents
DuPont™
TY198TWH2X0025PI
2X-Large
Case of 25 Each for $341.59
18-888-823
View Documents
DuPont™
TY198TWH3X0025PI
3X-Large
Case of 25 Each for $362.21
18-888-824
View Documents
DuPont™
TY198TWH4X0025PI
4X-Large
Case of 25 Each for $391.14
18-888-825
View Documents
DuPont™
TY198TWH5X0025PI
5X-Large
Case of 25 Each for $422.71
18-888-826
View Documents
DuPont™
TY198TWH6X0025PI
6X-Large
Case of 25 Each for $456.40
18-888-827
View Documents
DuPont™
TY198TWH7X0025PI
7X-Large
Case of 25 Each for $492.93
18888829
View Documents
DuPont™
TY198TWHMD0025PI
Medium
Case of 25 for $322.36
18888830
View Documents
DuPont™
TY198TWHSM0025PI
Small
Case of 25 for $322.36
18888831
View Documents
DuPont™
TY198TWHXL0025PI
X-Large
Case of 25 Each for $322.36
Description
Description
TYVEK™ 600 Coverall coveralls are ideal for occupations that require effective whole suit protection offered by a Type 4 suit because they are exposed to heavy liquid aerosols or toxic airborne solid particulates.
Enhanced design for increased flexibility and comfort:
  • Improved garment fit: extra flexibility and freedom of movement
  • New hood design: optimized to fit the contours of the face, ensuring fewer leakage points during head movement
  • Larger zipper puller: much easier to grasp when wearing gloves.
  • Certified to latest European Standards: including the Type 4 spray test.
Certifications
Chemical Protective Clothing, Cat.III,Type 5-B,6-B
Specifications
Specifications
Unisex
White
Elastic
Hooded
Non-sterile
Chemical Protective Clothing, Cat.III,Type 5-B,6-B
Yes
25/Cs.
Tyvek
Single-use
Elastic
Taped
Liquid, Particle
Packaged Individually
None
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'