DuPont™ Tyvek™ 600 Coverall with Hood, Elastic Ankles, Thumbloops
DuPont™ Tyvek™ 600 Coverall with Hood provides an effective barrier against low concentration water-based inorganic chemicals, small sized hazardous particles down to one micron, and biological hazards.
Supplier:  DuPont™ TY198TWH6X0025PI
 View more versions of this product
Catalog No. 18-888-826
Explore available promotions
Description
Description
TYVEK™ 600 Coverall coveralls are ideal for occupations that require effective whole suit protection offered by a Type 4 suit because they are exposed to heavy liquid aerosols or toxic airborne solid particulates.
Enhanced design for increased flexibility and comfort:
  • Improved garment fit: extra flexibility and freedom of movement
  • New hood design: optimized to fit the contours of the face, ensuring fewer leakage points during head movement
  • Larger zipper puller: much easier to grasp when wearing gloves.
  • Certified to latest European Standards: including the Type 4 spray test.
Certifications
Chemical Protective Clothing, Cat.III,Type 5-B,6-B
Specifications
Specifications
6X-Large
Tyvek
Single-use
Elastic
Taped
Liquid, Particle
Packaged Individually
None
Unisex
White
Elastic
Hooded
Non-sterile
Chemical Protective Clothing, Cat.III,Type 5-B,6-B
Yes
25/Cs.