missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DuPont™ Tyvek™ IsoClean™ Front Snap Cleanroom Frocks, High Mandarin Collar

Cleanroom frocks feature a high mandarin collar with snap for a secure fit and provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.

Supplier:  DuPont™ IC263SWHLG00300B

 View more versions of this product

Catalog No. 17-988-075


Explore available promotions

Alert:

Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.


Description

Description

Barrier to dry particulates, nonhazardous liquids, and microorganisms

  • Snap closure (6 snaps)
  • Elastic wrists
  • Color: White
  • Comfortable, lightweight, and durable
  • Bulk packaged in double transparent poly liners
Specifications

Specifications

Controlled Environments Frock
Tyvek
Single-use
Snaps
Serged
36 to 38 in.
39.5 in.
36.5 in.
Class 1000 (ISO 6)
Large
White
Non-sterile
Elastic
High Mandarin
Set-in Sleeve
100.33 cm
92.71 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions