missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DuPont™ Tyvek™ IsoClean™ Front Snap Cleanroom Frocks, High Mandarin Collar

Cleanroom frocks feature a high mandarin collar with snap for a secure fit and provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.

$255.42 - $292.40

Specifications

Product Type Controlled Environments Frock
Material Tyvek
Color White
Disposable Single-use
Sterility Non-sterile
View More Specs

Alert:

Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.


Products 7
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-988-073
View Documents
DuPont™
IC263SWHSM00300B
Small
Case of 30 Each for $255.42
 
17-988-074
View Documents
DuPont™
IC263SWHMD00300B
Medium
Case of 30 Each for $255.42
 
17-988-075
View Documents
DuPont™
IC263SWHLG00300B
Large
Case of 30 Each for $255.42
 
17-988-076
View Documents
DuPont™
IC263SWHXL00300B
X-Large
Case of 30 Each for $255.42
 
17-988-077
View Documents
DuPont™
IC263SWH2X00300B
2X-Large
Case of 30 Each for $255.42
 
17988078
View Documents
DuPont™
IC263SWH3X00300B
3X-Large
Case of 30 Each for $270.70
 
17988079
View Documents
DuPont™
IC263SWH4X00300B
4X-Large
Case of 30 Each for $292.40
 
Description

Description

Barrier to dry particulates, nonhazardous liquids, and microorganisms

  • Snap closure (6 snaps)
  • Elastic wrists
  • Color: White
  • Comfortable, lightweight, and durable
  • Bulk packaged in double transparent poly liners
Specifications

Specifications

Controlled Environments Frock
White
Non-sterile
Elastic
High Mandarin
Class 1000 (ISO 6)
Tyvek
Single-use
Snaps
Serged
Set-in Sleeve
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'