3M™ Ultimate FX FF-400 Series Full Facepiece Respirator
Rugged facepiece equipped with a large Scotchgard™ coated lens is designed to provide flexibility in extreme environments.
Supplier:  3M™ 7000029940
 View more versions of this product
Catalog No. 19-177-855
Explore available promotions
Description
Description
  • Large lens with peripheral vision delivers maximum comfort with a wide field of view
  • Exclusive Scotchgard™ coating on lens causes some liquids to bead up on the surface so they can be wiped off easily
  • Durable, permanently attached gaskets for easy maintenance and longer life
  • Low-profile bayonet design improves balance and offers better weight distribution
  • Durable head harness with six-strap configuration fits securely
  • Comfort cradle positions respirator comfortably on head
  • Soft, silicone nose cup and faceseal fits broad range of face shapes and sizes
  • Cool Flow™ exhalation valve allows easier breathing, reduces heat and moisture buildup
Compliance
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
Specifications
Bayonet
Silicone
Small
Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding
Speaking diaphragm
FF-401
Black
Full Facepiece Respirator
Silicone
3M™ Particle Filters 2000, 2200, 7000, or 5000 Series, 3M™ 6000 Series Gas/Vapor Cartridges and 3M™ 6000 Combination Cartridges
Scotchgard™
6-point