3M™ Ultimate FX FF-400 Series Full Facepiece Respirator
Rugged facepiece equipped with a large Scotchgard™ coated lens is designed to provide flexibility in extreme environments.
Supplier: 3M™ 7000002284
Description
- Large lens with peripheral vision delivers maximum comfort with a wide field of view
- Exclusive Scotchgard™ coating on lens causes some liquids to bead up on the surface so they can be wiped off easily
- Durable, permanently attached gaskets for easy maintenance and longer life
- Low-profile bayonet design improves balance and offers better weight distribution
- Durable head harness with six-strap configuration fits securely
- Comfort cradle positions respirator comfortably on head
- Soft, silicone nose cup and faceseal fits broad range of face shapes and sizes
- Cool Flow™ exhalation valve allows easier breathing, reduces heat and moisture buildup
Compliance
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
Bayonet | |
Silicone | |
Medium | |
Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding | |
Speaking diaphragm | |
FF-402 |
Black | |
Full Facepiece Respirator | |
Silicone | |
3M™ Particle Filters 2000, 2200, 7000, or 5000 Series, 3M™ 6000 Series Gas/Vapor Cartridges and 3M™ 6000 Combination Cartridges | |
Scotchgard™ | |
6-point |