missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Undecafluorohexanoic Acid High Grade 98.0+%, TCI America™

$160.90 - $477.51

Chemical Identifiers

CAS 307-24-4
Molecular Formula C6HF11O2
Molecular Weight (g/mol) 314.054
MDL Number MFCD00198040
InChI Key PXUULQAPEKKVAH-UHFFFAOYSA-N
Synonym Perfluorohexanoic Acid High Grade, IPC-PFFA-6 HG
PubChem CID 67542
ChEBI CHEBI:83492
IUPAC Name 2,2,3,3,4,4,5,5,6,6,6-undecafluorohexanoic acid
SMILES C(=O)(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A57221G
View Documents
TCI America
A57221G
1 g
Each for $160.90
 
A57225G
View Documents
TCI America
A57225G
5 g
Each for $477.51
 
Description

Description

Specifications

Chemical Identifiers

307-24-4
314.054
PXUULQAPEKKVAH-UHFFFAOYSA-N
67542
2,2,3,3,4,4,5,5,6,6,6-undecafluorohexanoic acid
C6HF11O2
MFCD00198040
Perfluorohexanoic Acid High Grade, IPC-PFFA-6 HG
CHEBI:83492
C(=O)(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

307-24-4
Colorless
C6HF11O2
1 g
Perfluorohexanoic Acid High Grade, IPC-PFFA-6 HG
C(=O)(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)O
314.054
CHEBI:83492
≥98.0% (T)
Undecafluorohexanoic Acid High Grade [Ion-Pair Reagent for LC-MS]
14°C
157°C
MFCD00198040
3265
PXUULQAPEKKVAH-UHFFFAOYSA-N
2,2,3,3,4,4,5,5,6,6,6-undecafluorohexanoic acid
67542
314.05
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1182

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'