Red Kap™ Unisex White Specialized Cuffed Lab Coats
Unisex poly cotton specialized lab coat with exterior pockets and knit cuffs features a 6 gripper secure closure.
$49.84 - $49.84
Specifications
Product Type General Purpose Lab Coat
Gender Unisex
Material Polyester, Cotton
Color White
Closure Type Snaps
View More Specs
Products 5
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
19080340
View Documents
Red Kap™
KP70WHRGXS
Each for $49.84
19080341
View Documents
Red Kap™
KP70WHRGS
Each for $49.84
19080342
View Documents
Red Kap™
KP70WHRGM
Each for $49.84
19080343
View Documents
Red Kap™
KP70WHRGL
Each for $49.84
19080344
View Documents
Red Kap™
KP70WHRGXL
Each for $49.84
Description
Description
  • Chest pocket and two lower pockets for convenient storage
  • Knit cuffs that keep sleeves from interfering with work
  • Extended back coverage
  • Primary closure is snap, gripper
  • Wash care is industrial laundry for heavy soil and home wash for light soil
  • Fabric is 5 oz. performance blend poplin
  • 80% Polyester, 20% Combed Cotton
  • Wrinkle-resistant finish
  • Six gripper closure
  • One-piece, lined, notched lapel collar
  • Knit wristlet cuffs
  • Requires minimum ironing
  • No side vent openings
  • Unisex, meant to be worn over clothes
Specifications
Specifications
General Purpose Lab Coat
Polyester, Cotton
Snaps
Reusable
Standard Length
Not Autoclavable
Unisex
White
Knit
Non-sterile
3
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'