Thermo Scientific™ Universal Test Tube Racks with Holder for MaxQ™ High Performance Shakers
Shake multiple tubes at an adjustable angle to maximize sample aeration. The rack holders are adjustable to fit any of the four available test tube rack sizes.
Supplier:  Thermo Scientific™ 950060
 View more versions of this product
Catalog No. 11676043
Description
Description
Shake multiple tubes at an adjustable angle to maximize sample aeration. Thermo Scientific™ Universal Test Tube Racks with Holder for MaxQ™ 8000 Shakers provide ultimate flexibility—they can be adjusted to fit any of the four available test tube rack sizes.
  • Universal test tube rack holder can be adjusted to fit any of the four available test tube rack sizes
  • Universal design requires the separate purchase of a test tube rack
Specifications
Specifications
Tube
10 mL, 20 mL
Test tube rack
40
16 to 20 mm
Test Tube Rack
MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000