Idex PEEK Chromatography Tubing
An excellent replacement for stainless steel tubing
Supplier: Idex UP1531BXL
Description
An excellent replacement for stainless steel tubing
Can be used in place of stainless steel tubing in most liquid analytical systems
- Biocompatible, inert and easy to cut
- Great for high-pressure applications
- Wide choice of outer and inner diameters
- Many sizes color-coded for easier inner diameter identification
Specifications
Tubing | |
High-Pressure Applications | |
100 ft. |
Blue | |
0.01 in. | |
7000 psi |