Idex PEEK Chromatography Tubing DFS Item
An excellent replacement for stainless steel tubing
$83.75 - $1342.53
Specifications
For Use With (Application) High-Pressure Applications
View More Specs
Products 16
Catalog Number Mfr. No. Color Diameter (English) Inner Overall Length (English) Maximum Pressure Price Quantity  
Catalog Number Mfr. No. Color Diameter (English) Inner Overall Length (English) Maximum Pressure Price Quantity  
05-701 DFS Item
View Documents
Idex
UP1535DUP
Red 0.005 in. 5 ft. 7000 psi (483 bar)
Each for $118.53
05-701-10 DFS Item
View Documents
Idex
UP1531L
Natural 0.01 in. 50 ft. 7000 psi
Each for $552.87
05-701-1 DFS Item
View Documents
Idex
UP1536
Yellow 0.007 in. 5 ft. 7000 psi
Each for $83.75
05-701-13 DFS Item
View Documents
Idex
UP1531BXL
Blue 0.01 in. 100 ft. 7000 psi
Each for $1,010.74
05-701-15 DFS Item
View Documents
Idex
UP1532XL
Orange 0.02 in. 100 ft. 7000 psi
Each for $1,010.74
05-701-18 DFS Item
View Documents
Idex
UP1535L
Red 0.005 in. 50 ft. 7000 psi
Each for $1,227.91
05-701-2 DFS Item
View Documents
Idex
UP1531
Natural 0.01 in. 5 ft. 7000 psi (483 bar)
Each for $83.75
05-701-20 DFS Item
View Documents
Idex
UP1536L
Yellow 0.007 in. 50 ft. 7000 psi (483 bar)
Each for $552.87
05-701-3 DFS Item
View Documents
Idex
UP1531BDUP
Blue 0.01 in. 5 ft. 7000 psi
Each for $83.75
05-701-4 DFS Item
View Documents
Idex
UP1532
Orange 0.02 in. 5 ft. 7000 psi (483 bar)
Each for $83.75
05-701-5 DFS Item
View Documents
Idex
UP1533DUP
Green 0.03 in. 5 ft. 7000 psi (483 bar)
Each for $88.19
05-701-6 DFS Item
View Documents
Idex
UP1538DUP
Natural 0.04 in. 5 ft. 5000 psi (345 bar)
Each for $83.75
05-701-8 DFS Item
View Documents
Idex
UP1534
Natural 0.062 in. 5 ft. 4000 psi
Each for $173.23
05-701-9 DFS Item
View Documents
Idex
UP1544
Natural 0.08 in. 5 ft. 3000 psi
Each for $199.38
05-701-19 DFS Item
View Documents
Idex
UP1535XL
Natural 0.005 in. 100 ft. 7000 psi
Each for $1,223.62
05-701-17 DFS Item
View Documents
Idex
1533XL
Green 0.03 in. 100 ft. 7000 psi
Each for $1,342.53
Description
Description
An excellent replacement for stainless steel tubing
Can be used in place of stainless steel tubing in most liquid analytical systems
  • Biocompatible, inert and easy to cut
  • Great for high-pressure applications
  • Wide choice of outer and inner diameters
  • Many sizes color-coded for easier inner diameter identification
Specifications
Specifications
High-Pressure Applications
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'