Idex PEEK Chromatography Tubing DFS Item
An excellent replacement for stainless steel tubing
Supplier:  Idex UP1531
 View more versions of this product
Catalog No. 05-701-2
Explore available promotions
Description
Description
An excellent replacement for stainless steel tubing
Can be used in place of stainless steel tubing in most liquid analytical systems
  • Biocompatible, inert and easy to cut
  • Great for high-pressure applications
  • Wide choice of outer and inner diameters
  • Many sizes color-coded for easier inner diameter identification
Specifications
Specifications
Tubing
High-Pressure Applications
5 ft.
7000 psi (483 bar)
Natural
0.01 in.
PEEK