Idex PEEK Chromatography Tubing
An excellent replacement for stainless steel tubing
Supplier: Idex UP1536L
Description
An excellent replacement for stainless steel tubing
Can be used in place of stainless steel tubing in most liquid analytical systems
- Biocompatible, inert and easy to cut
- Great for high-pressure applications
- Wide choice of outer and inner diameters
- Many sizes color-coded for easier inner diameter identification
Specifications
Tubing | |
High-Pressure Applications | |
50 ft. | |
7000 psi (483 bar) |
Yellow | |
0.007 in. | |
PEEK |