missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Idex Stainless-Steel Tubing 0.020 in.OD 0.005 in.ID DFS Item

Seamless, precut stainless steel tubing is specially prepared for the exacting requirements of todays analyses

Supplier:  Idex UPU117

 View more versions of this product

Catalog No. 05-701-108


Explore available promotions

Description

Description

Stainless steel is naturally self-passivating, forming an oxidation layer on newly created surfaces. Idex Heath & Science Scientific takes extra steps to ensure the chemical resistance of the stainless steel tubing by manually passivating before and after the tubing is cut into specified lengths.

  • Precut 316 stainless steel tubing:
  • Ends are machine cut and polished
  • Edges are deburred inside and outside
  • Tubing is passivated
  • Inside is flushed with reagent-grade isopropanol
  • Color-coded banding for easy identification of the inner diameter
  • All stainless-steel tubing longer than 1m is coiled

Specifications

Specifications

Tubing
1/16 in.
0.75 mm
20 cm
12,000 psi (827 bar)
White
0.030 in.
8 in.
Stainless Steel (316)
Videos
SDS
Documents

Documents

Promotions

Promotions