Idex Stainless-Steel Tubing 0.020 in.OD 0.005 in.ID
Seamless, precut stainless steel tubing is specially prepared for the exacting requirements of todays analyses
$12.13 - $1205.85
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Overall Length (Metric) | Material | Price | Qty | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Overall Length (Metric) | Material | Price | Qty | |||
05-701-106
![]() |
Idex
UPU115 |
0.75 mm | 5 cm | Stainless Steel (316) |
Each for $12.40
|
|
|||
05-701-108
![]() |
Idex
UPU117 |
0.75 mm | 20 cm | Stainless Steel (316) |
Each for $20.08
|
|
|||
05-701-109
![]() |
Idex
UPU118 |
0.75 mm | 30 cm | Stainless Steel (316) |
Each for $31.13
|
|
|||
05-701-119
![]() |
Idex
UPU128 |
0.175 mm | 20 cm | Stainless Steel (316) |
Each for $22.60
|
|
|||
05-701-126
![]() |
Idex
UPU135 |
0.5 mm | 1 m | Stainless Steel (304) |
Each for $68.26
|
|
|||
05-701-127
![]() |
Idex
UPU136 |
0.75 mm | 0.5 m | Stainless Steel (316) |
Each for $45.11
|
|
|||
05-701-128
![]() |
Idex
UPU137 |
0.75 mm | 1 m | Stainless Steel (316) |
Each for $79.75
|
|
|||
05-701-129
![]() |
Idex
UPU138 |
1.0 mm | 5 cm | Stainless Steel (316) |
Each for $17.00
|
|
|||
05-701-130
![]() |
Idex
UPU139 |
1.0 mm | 10 cm | Stainless Steel (316) |
Each for $26.08
|
|
|||
05-701-138
![]() |
Idex
UPU147 |
1.15 mm | 20 cm | Stainless Steel (316) |
Each for $68.70
|
|
|||
05-701-94
![]() |
Idex
UPU101 |
0.5 mm | 5 cm | Stainless Steel (304) |
Each for $12.13
|
|
|||
05-701-95
![]() |
Idex
UPU102 |
0.5 mm | 10 cm | Stainless Steel (304) |
Each for $14.68
|
|
|||
05-701-96
![]() |
Idex
UPU103 |
0.5 mm | 20 cm | Stainless Steel (304) |
Each for $21.60
|
|
|||
05-701-97
![]() |
Idex
UPU104 |
0.5 mm | 30 cm | Stainless Steel (304) |
Each for $24.92
|
|
|||
29-008-954
![]() |
Idex
UPU800 |
2.0 mm | 1 m | Stainless Steel (316) |
Each for $246.21
|
|
|||
05-701-141
![]() |
Idex
U150** |
1.15 mm | 1 m | Stainless Steel (316) |
Each for $175.08
|
|
|||
05-701-179
![]() |
Idex
UPU805 |
2.0 mm | 5 m | Stainless Steel (316) |
Each for $1,205.85
|
|
|||
05-701-104
![]() |
Idex
UPU113* |
0.25 mm | 20 cm | Stainless Steel (316) |
Each for $21.21
|
|
|||
05-701-102
![]() |
Idex
UPU111 |
0.25 mm | 5 cm | Stainless Steel (316) |
Each for $12.77
|
|
|||
05-701-100
![]() |
Idex
UPU107* |
0.75 mm | 1.5 m | Stainless Steel (316) |
Each for $104.77
|
|
|||
05-701-117
![]() |
Idex
UPU126 |
0.175 mm | 5 cm | Stainless Steel (316) |
Each for $12.51
|
|
|||
05-701-118
![]() |
Idex
UPU127 |
0.175 mm | 10 cm | Stainless Steel (316) |
Each for $16.19
|
|
|||
05-701-120
![]() |
Idex
UPU129 |
0.175 mm | 30 cm | Stainless Steel (316) |
Each for $80.74
|
|
|||
05-701-124
![]() |
Idex
UPU133 |
0.25 mm | 1 m | Stainless Steel (316) |
Each for $121.12
|
|
|||
05-701-121
![]() |
Idex
UPU130 |
0.175 mm | 0.5 m | Stainless Steel (316) |
Each for $45.25
|
|
|||
05-701-147
![]() |
Idex
UPU156 |
0.125 mm | 0.5 m | Stainless Steel (316) |
Each for $49.72
|
|
|||
05-701-99
![]() |
Idex
UPU106 |
0.25 mm | 1.5 m | Stainless Steel (316) |
Each for $99.66
|
|
|||
Description
Stainless steel is naturally self-passivating, forming an oxidation layer on newly created surfaces. Idex Heath & Science Scientific takes extra steps to ensure the chemical resistance of the stainless steel tubing by manually passivating before and after the tubing is cut into specified lengths.
- Precut 316 stainless steel tubing:
- Ends are machine cut and polished
- Edges are deburred inside and outside
- Tubing is passivated
- Inside is flushed with reagent-grade isopropanol
- Color-coded banding for easy identification of the inner diameter
- All stainless-steel tubing longer than 1m is coiled
Specifications
Tubing |