missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Idex Stainless-Steel Tubing 0.020 in.OD 0.005 in.ID
Seamless, precut stainless steel tubing is specially prepared for the exacting requirements of todays analyses
Supplier: Idex UPU128
Description
Stainless steel is naturally self-passivating, forming an oxidation layer on newly created surfaces. Idex Heath & Science Scientific takes extra steps to ensure the chemical resistance of the stainless steel tubing by manually passivating before and after the tubing is cut into specified lengths.
- Precut 316 stainless steel tubing:
- Ends are machine cut and polished
- Edges are deburred inside and outside
- Tubing is passivated
- Inside is flushed with reagent-grade isopropanol
- Color-coded banding for easy identification of the inner diameter
- All stainless-steel tubing longer than 1m is coiled
Specifications
Tubing | |
1/16 in. | |
0.175 mm | |
20 cm | |
20,900 psi (1.441 bar) |
Black | |
0.007 in. | |
8 in. | |
Stainless Steel (316) |