missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Uranine (Concentrated, Water Soluble/Laboratory), Fisher Chemical™

Used by nuclear utilities to test cooling systems for small leaks

Supplier:  Thermo Fisher Scientific FLA833100

 View more versions of this product

Catalog No. A833-100



Explore available promotions

Description

Description

Specifications

Specifications

Uranine
C20H10Na2O5
Fluorescein Sodium Salt
NJDNXYGOVLYJHP-UHFFFAOYSA-L
disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
376.28
Glass Bottle
100 g
320°C
MFCD00167039
Pass Test
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
376.28
Laboratory
Orange
Powder

Chemical Identifiers

C20H10Na2O5
MFCD00167039
Fluorescein Sodium Salt
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
376.28
NJDNXYGOVLYJHP-UHFFFAOYSA-L
disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Videos
Safety and Handling

Safety and Handling

SDS Documents

Documents

Promotions

Promotions