missing translation for 'onlineSavingsMsg'
Learn More

Uranine (Concentrated, Water Soluble/Laboratory), Fisher Chemical™

Used by nuclear utilities to test cooling systems for small leaks

$161.27 - $391.17

Chemical Identifiers

CAS 518-47-8
Molecular Formula C20H10Na2O5
Molecular Weight (g/mol) 376.28
MDL Number MFCD00167039
InChI Key NJDNXYGOVLYJHP-UHFFFAOYSA-L
Synonym Fluorescein Sodium Salt
IUPAC Name disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
A833-100
SDS View Documents Promotion Details
Thermo Fisher Scientific
FLA833100
100 g Glass Bottle
Each for $161.27
Case of 6 Each for $967.62
Only null left
 
A833-500
SDS View Documents Promotion Details
Fisher Chemical
FLA833500
500 g Poly Bottle
Each for $391.17
Case of 6 Each for $2,347.02
Only null left
 
Description

Description

Specifications

Chemical Identifiers

518-47-8
376.28
NJDNXYGOVLYJHP-UHFFFAOYSA-L
disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
C20H10Na2O5
MFCD00167039
Fluorescein Sodium Salt
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12

Specifications

320°C
MFCD00167039
Pass Test
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
376.28
Laboratory
Orange
Powder
C20H10Na2O5
Fluorescein Sodium Salt
NJDNXYGOVLYJHP-UHFFFAOYSA-L
disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
376.28
Glass Bottle
100 g
Uranine
Videos
Safety and Handling

Safety and Handling

SDS
missing translation for 'documents'

missing translation for 'documents'