missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Urea, ≥98%, Molecular Biology Reagent Grade, MP Biomedicals™
Supplier: MP Biomedicals Inc 0219485790
Specifications
Urea | |
White | |
≥98% | |
MFCD00008022 | |
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate | |
C(=O)(N)N | |
60.056 | |
CHEBI:48376 | |
Molecular Biology Reagent |
57-13-6 | |
1.335g/mL at 25°C (Lit.) | |
CH4N2O | |
500 g | |
XSQUKJJJFZCRTK-UHFFFAOYSA-N | |
urea | |
1176 | |
60.06 | |
Powder |
Chemical Identifiers
57-13-6 | |
60.056 | |
XSQUKJJJFZCRTK-UHFFFAOYSA-N | |
1176 | |
urea |
CH4N2O | |
MFCD00008022 | |
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate | |
CHEBI:48376 | |
C(=O)(N)N |
Safety and Handling
EINECSNumber : 200-315-5
Recommended Storage : Store at Room Temperature (15 to 30°C).